Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerebellin

Chemical Identifiers

Chemical Name or Material Cerebellin
CAS 94071-26-8
Formula Weight 1632.8
MDL Number MFCD00167553
Synonym Ser-Gly-Ser-Ala-Lys-Val-Ala-Phe-Ser-Ala-Ile-Arg-Ser-Thr-Asn-His
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66631MCR

  SDS
alfa aesar™
J66631MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1632.8
Chemical Name or Material Cerebellin
MDL Number MFCD00167553
Physical Form Solid
Storage Note 1 -30° to -10°C
Color White to off-white
Quantity 1mg
CAS 94071-26-8
Synonym Ser-Gly-Ser-Ala-Lys-Val-Ala-Phe-Ser-Ala-Ile-Arg-Ser-Thr-Asn-His
Safety and Handling

-30° to -10°C