Please login to your online account to display your discounted pricing

Ceramide, Tocris


Ser/Thr protein phosphatase activator

Quantity 10mg

Manufacturer: r & d systems  RDS074410

 View more versions of this product

Catalog No. 074410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 341.53
Assay Percent Range >98%
Molecular Formula C20H39NO3
CAS 3102-57-6
Chemical Name or Material Ceramide
Synonym N-Acetylsphingosine
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Ceramide
Assay Percent Range >98%
Molecular Formula C20H39NO3
Formula Weight 341.53
Synonym N-Acetylsphingosine
Accessories

Desiccate at -20°C