missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cephalothin sodium salt, 50mg/mL in distilled water, sterile-filtered, Thermo Scientific™
$93.74 - $93.74
Chemical Identifiers
CAS | 58-71-9 |
---|---|
Molecular Formula | C16H15N2NaO6S2 |
Molecular Weight (g/mol) | 418.414 |
MDL Number | MFCD00072025 |
InChI Key | VUFGUVLLDPOSBC-XRZFDKQNSA-M |
PubChem CID | 23675321 |
ChEBI | CHEBI:3542 |
IUPAC Name | sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
SMILES | CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67256XF
|
Thermo Scientific Chemicals
J67256XF |
1 mL |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
58-71-9 | |
418.414 | |
VUFGUVLLDPOSBC-XRZFDKQNSA-M | |
CHEBI:3542 | |
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+] |
C16H15N2NaO6S2 | |
MFCD00072025 | |
23675321 | |
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
Specifications
Cephalothin Sodium Salt | |
58-71-9 | |
MFCD00072025 | |
CC(=O)OCC1=C(N2C(C(C2=O)NC(=O)CC3=CC=CS3)SC1)C(=O)[O-].[Na+] | |
418.414 | |
CHEBI:3542 | |
Cephalothin sodium salt, 50mg/mL in distilled water, sterile-filtered |
Liquid | |
C16H15N2NaO6S2 | |
VUFGUVLLDPOSBC-XRZFDKQNSA-M | |
sodium;(6R,7R)-3-(acetyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
23675321 | |
1 mL |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334-H335
EINECSNumber : 200-394-6
RTECSNumber : XI0388300
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C