Please login to your online account to display your discounted pricing

Cephalexin hydrate, 99.23%, MP Biomedicals™


Quantity 25g

Manufacturer: mp biomedicals  19866625

 View more versions of this product

Catalog No. ICN19866625

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 0.9923
Physical Form Powder
Formula Weight 365.4
Color White
Quantity 25g
Molecular Formula C16H17N3O4S·H2O
CAS 15686-71-2
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid
Chemical Name or Material Cephalexin hydrate

Specifications

Chemical Name or Material Cephalexin hydrate
CAS 15686-71-2
Physical Form Powder
Molecular Formula C16H17N3O4S·H2O
Formula Weight 365.4
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid