Please login to your online account to display your discounted pricing

Cephalexin hydrate, 99.23%, MP Biomedicals™

$574.90 - $574.90

Chemical Identifiers

Chemical Name or Material Cephalexin hydrate
CAS 15686-71-2
Physical Form Powder
Molecular Formula C16H17N3O4S·H2O
Formula Weight 365.4
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19866625

 
mp biomedicals
19866625
25g Each for $574.90
Description & Specifications

Specifications

Physical Form Powder
Assay 0.9923
Formula Weight 365.4
Color White
Quantity 25g
Molecular Formula C16H17N3O4S·H2O
Chemical Name or Material Cephalexin hydrate
CAS 15686-71-2
Synonym 7-(d-a-Aminophenylacetamido)desacetoxycephalosporanic acid