Learn More
Cephalexin hydrate, 97+%, Thermo Scientific Chemicals
Cephalexin hydrate, cefalexin hydrate, CAS # 23325-78-2, is a first-generation cephalosporin antibiotic from the beta-lactam family.
$137.27 - $501.19
Chemical Identifiers
CAS | 15686-71-2 |
---|---|
Molecular Formula | C16H20ClN3O5S |
Molecular Weight (g/mol) | 401.862 |
MDL Number | MFCD00167148 |
InChI Key | YHJDZIQOCSDIQU-OEDJVVDHSA-N |
PubChem CID | 57515946 |
IUPAC Name | (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate |
SMILES | [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6317206
|
Thermo Scientific Chemicals
J6317206 |
5 g |
N/A
|
|
|||||
AAJ6317214
|
Thermo Scientific Chemicals
J6317214 |
25 g |
N/A
|
|
|||||
Description
Cephalexin is a cephalosporin antibiotic used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis. Cephalexin disrupts the synthesis of the peptidoglycan layer of bacterial cell walls which is responsible for cell wall structural integrity. Peptidoglycan synthesis is facilitated by transpeptidases known as penicillin-binding proteins (PBPs). PBPs bind to the D-Ala-D-Ala at the end of muropeptides (peptidoglycan precursors) to crosslink the peptidoglycan. Cephalexin antibiotics mimic the D-Ala-D-Ala site, thereby competitively inhibiting PBP crosslinking of peptidoglycan.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Cephalexin hydrate is the hydrate of the cephalosporin antibiotic, cephalexin
• Cephalexin is a beta-lactam antibiotic that inhibits bacterial growth and division
• Cephalexin binds to penicillin-binding protein and inhibits cell wall synthesis
Application
• Cephalexin works best on gram-positive microbes
• Useful for immunology research, particularly on antibiotic allergies and cross-reactivity
Chemical Identifiers
15686-71-2 | |
401.862 | |
YHJDZIQOCSDIQU-OEDJVVDHSA-N | |
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate |
C16H20ClN3O5S | |
MFCD00167148 | |
57515946 | |
[H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-] |
Specifications
Cephalexin Hydrate | |
15686-71-2 | |
MFCD00167148 | |
141974 | |
YHJDZIQOCSDIQU-OEDJVVDHSA-N | |
(6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate | |
57515946 | |
≥97% | |
5 g |
Powder | |
C16H20ClN3O5S | |
965503 | |
Soluble in DMSO and Methanol. | |
[H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-] | |
401.862 | |
347.39 (Anhydrous) | |
White | |
Cephalexin hydrate |
Safety and Handling
May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction
Respiratory sensitisation (category 1), Skin sensitisation (category 1)
EINECSNumber : 239-773-6
RTECSNumber : XI0350000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only