Please login to your online account to display your discounted pricing

Alfa Aesar™ Cephalexin, 50mg/mL in 1M ammonium hydroxide, sterile-filtered

Chemical Identifiers

Chemical Name or Material Cephalexin
CAS 15686-71-2
Name Note 50mg/mL in 1M ammonium hydroxide, sterile-filtered
MDL Number MFCD00167148
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67377AC

 
alfa aesar™
J67377AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67377AD

 
alfa aesar™
J67377AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Cephalexin
MDL Number MFCD00167148
CAS 15686-71-2
Health Hazard 3 GHS P Statement
P285-P261-P280-P305+P351+P338-P310-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H318-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
Storage Note 1 -30° to -10°C
Name Note 50mg/mL in 1M ammonium hydroxide, sterile-filtered
Quantity 25mL
RTECS Number XI0350000
Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P305+P351+P338-P310-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H318-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.

-30° to -10°C