CEP Poly-Stacker and Poly-Racker
Manufacturer:  CEP 6002YE
 View more versions of this product
Catalog No. 191202554
Description
Description
Specifications
Specifications
Yellow
Manufacturing plants, mining, chemical storage facilities
13 in.
Two 55 gal. drums
124.46cm
1088.6kg
25.4kg
104.14cm
Poly-Stacker™ Drum Rack
49 x 41 x 13 in. (124.46 x 104.14 x 33cm)
Poly-Stacker™
33cm
49 in.
2400 lb.
56 lb.
41 in.
Polyethylene