CEP Poly Overpack™ 20 and 30 Drums
Directly contain solids and sludges with security
Manufacturer:  CEP 1230YE
 View more versions of this product
Catalog No. 19120558
Description
Description
  • With one-half turn push-down lid allows easy access, unlike traditional “hoop-ring” designs
  • Ergonomically designed units accept bottles, cans, pails, and drums
  • Tough HDPE polyethylene construction is weather and chemical resistant
  • Built-in shoulder for easy handling; nestable for ease of shipment and storage
  • Wide mouth, rigid walls, small footprint and integral handles make these drums perfect for overpacking damaged containers and for holding spill kits
Compliance
Meet performance-oriented packaging requirements of U.S. DOT 49CFR173.3(c) and UN 1H2/X100/S DOT 49CFR173.3(c).
Specifications
Specifications
UN 1H2/X100/S DOT 49 CFR 173.3
Overpacking damaged containers, emergency response and medical and operating room waste destined for the freezer or incineration
72.39 to 76.2cm
221 lb.
13 lb.
HDPE
Yellow
28.5 to 30 in.
16 gal. Drums
100.24kg
5.9kg
Poly Overpack