CEP Oil Only Booms, Pillows and Socks
Variety of sorbents for all types of oil spills
Supplier: CEP SAKPIL10
Description
- Hydrophobic, hydrophilic, static resistant
- Nonlinting or anti-shed netting
- Overlapping clips for deployment in any length
- Pillows and socks are ideal for catch basins and sumps
Specifications
White | |
158.9 L | |
Sorbent Sack Pillow | |
25 x 14 in. (63.5 x 35.5 cm) | |
63.5 cm | |
35.5 cm | |
35.5 cm |
42 gal. | |
Polypropylene | |
Oil spill control, spill containment, oil spill clean up and skimming oil from water | |
25 in. | |
14 in. | |
14 in. |