CEP Oil Only Booms, Pillows and Socks
Variety of sorbents for all types of oil spills
$146.66 - $226.94
Specifications
Color White
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Type Diameter (English) Price Quantity  
Catalog Number Mfr. No. Type Diameter (English) Price Quantity  
29-635-3054
View Documents Promotion Details
CEP
SAKPIL10
Sorbent Sack Pillow 14 in.
Case of 10 Each for $146.66
29-636-4235
View Documents Promotion Details
CEP
B510
Sorbent Boom 5 in.
Pack of 4 Each for $226.94
Description
Description
  • Hydrophobic, hydrophilic, static resistant
  • Nonlinting or anti-shed netting
  • Overlapping clips for deployment in any length
  • Pillows and socks are ideal for catch basins and sumps
Specifications
Specifications
White
Polypropylene