CEP Oil Only Booms, Pillows and Socks
Variety of sorbents for all types of oil spills
Manufacturer:  CEP B510
 View more versions of this product
Catalog No. 29-636-4235
Description
Description
  • Hydrophobic, hydrophilic, static resistant
  • Nonlinting or anti-shed netting
  • Overlapping clips for deployment in any length
  • Pillows and socks are ideal for catch basins and sumps
Specifications
Specifications
White
12.7 cm
Sock
121.1 L
3.04 m
Sorbent Boom
5 in.
Wastewater troughs, loading docks, ponds, rivers and oceans to absorb oily spills
32 gal.
10 ft.
Polypropylene