missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CEP Oil-Only Bonded Sorbents

Ideal for wiping applications

Manufacturer:  CEP BER144

 View more versions of this product

Catalog No. 29-636-4232



Description

Description

  • Sonic bonded to create stronger, more durable fiber without affecting adsorption
  • Fast-wicking fibers absorb up to 25 times their weight in hydrocarbons
  • Provide containment and absorption while repelling aquatic liquids

Specifications

Specifications

Polypropylene
76.2cm
39 gal.
45.7m
Bonded Sorbent Roll
Heavy
150 ft. x 30 in. (45.7m x 76.2cm)
30 in.
White
147.6L
150 ft.
Bonded for increased strength
39 gal., 147.6L
SDS
Documents