missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CEP Oil-Only Bonded Sorbents
Ideal for wiping applications
Manufacturer: CEP BEP100
Description
- Sonic bonded to create stronger, more durable fiber without affecting adsorption
- Fast-wicking fibers absorb up to 25 times their weight in hydrocarbons
- Provide containment and absorption while repelling aquatic liquids
Specifications
Polypropylene | |
38cm | |
44 gal. | |
43.18cm | |
Bonded Sorbent Pad | |
Medium | |
17 x 15 in. (43.18 x 38cm) |
15 in. | |
White | |
166.5L | |
17 in. | |
Bonded for increased strength | |
44 gal., 166.5L |