missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CEP Oil-Only Bonded Sorbents

Ideal for wiping applications

Manufacturer:  CEP BR144

 View more versions of this product

Catalog No. 19-052-058



Description

Description

  • Sonic bonded to create stronger, more durable fiber without affecting adsorption
  • Fast-wicking fibers absorb up to 25 times their weight in hydrocarbons
  • Provide containment and absorption while repelling aquatic liquids

Specifications

Specifications

White
150 ft. x 30 in. (45.7 m x 76.2 cm)
50 gal.
150 ft.
Polypropylene
30 in.
Bonded for increased strength
Medium-weight
189 L
45.7 m
Bonded Sorbent Roll
76.2 cm
SDS
Documents
Product Certifications