missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cellulose microcrystalline, extra pure, average particle size 90μm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 382310010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidSpecifications
White | |
5 to 7 | |
9004-34-6 | |
Solubility in water: insoluble. | |
Extra Pure | |
1.5% max. | |
250 Micron <1% min., 75 Micron >45% min., 32 Micron >70% min. |
Cellulose microcrystalline | |
Fine Crystalline Powder or Granular Powder | |
(C12H20O10)n | |
1 kg | |
Authentic | |
Glass bottle | |
0.05% max. |
Chemical Identifiers
9004-34-6 |
RUO – Research Use Only