Please login to your online account to display your discounted pricing

Cellulose Acetate Propionate, Average M.W. 25,000, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
MDL Number MFCD00146341
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC404282500

  SDS
thermo fisher scientific
AC404282500
Plastic bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Powder
Quantity 250g
Color Almost white
TSCA TSCA
Chemical Name or Material Cellulose acetate propionate
MDL Number MFCD00146341
CAS 9004-39-1
Infrared Spectrum Authentic
Moisture moisture (as packaged): < 3.0%
Packaging Plastic bottle
Propionyl Content 43 to 47% (GC)
Viscosity 0.2 to 0.6 (sec.)
Acetyl Content acetyl content (GC): 0.5 to 2.5%
Free Acid 0.06% max.