Please login to your online account to display your discounted pricing

Cellulose Acetate Propionate, Average M.W. 25,000, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC404282500

 View more versions of this product

Catalog No. AC404282500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Powder
Quantity 250g
Color Almost white
TSCA TSCA
Propionyl Content 43 to 47% (GC)
Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
MDL Number MFCD00146341
Infrared Spectrum Authentic
Viscosity 0.2 to 0.6 (sec.)
Packaging Plastic bottle
Moisture moisture (as packaged): < 3.0%
Acetyl Content acetyl content (GC): 0.5 to 2.5%
Free Acid 0.06% max.

Specifications

Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
MDL Number MFCD00146341