Please login to your online account to display your discounted pricing

Cellulose Acetate Propionate, Average M.W. 15,000, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
Formula Weight 15,000g/mol (Average )
MDL Number MFCD00146341
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC404275000

  SDS
thermo fisher scientific
404275000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Acetyl Content Acetyl content (GC): < 1.0%
Free Acid 0.04% max.
Hydroxyl Content Hydroxyl: 4.4 tot 5.6%
Infrared Spectrum Authentic
Moisture Moisture (as packaged): < 3.0%
Packaging Plastic bottle
Propionyl Content 40 to 45% (GC)
Viscosity 0.10 to 0.20 (sec.)
Color White
Quantity 500g
Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
Physical Form White fine powder
Formula Weight 15,000g/mol (Average )
MDL Number MFCD00146341
TSCA TSCA