Please login to your online account to display your discounted pricing

Cellulose Acetate Propionate, Average M.W. 15,000, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  404275000

 View more versions of this product

Catalog No. AC404275000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Acetyl Content Acetyl content (GC): < 1.0%
Free Acid 0.04% max.
Hydroxyl Content Hydroxyl: 4.4 tot 5.6%
Infrared Spectrum Authentic
Moisture Moisture (as packaged): < 3.0%
Packaging Plastic bottle
Propionyl Content 40 to 45% (GC)
Viscosity 0.10 to 0.20 (sec.)
Color White
Quantity 500g
Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
Physical Form White fine powder
Formula Weight 15,000g/mol (Average )
MDL Number MFCD00146341
TSCA TSCA

Specifications

Chemical Name or Material Cellulose acetate propionate
CAS 9004-39-1
Formula Weight 15,000g/mol (Average )
MDL Number MFCD00146341