Please login to your online account to display your discounted pricing

Cellulose Acetate Butyrate, Butyryl Content 50-54%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bucket

Manufacturer: thermo fisher scientific  AC404220025

 View more versions of this product

Catalog No. AC404220025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Granular Crystalline Powder
Quantity 2.5kg
Color White
TSCA TSCA
Name Note Butyryl Content 50-54%
Hydroxyl Content hydroxyl content: 1.5% (typically)
Packaging Plastic bucket
Butyryl Content butyryl content: 50 to 54%
Chemical Name or Material Cellulose acetate butyrate
CAS 9004-36-8
MDL Number MFCD00081513
Infrared Spectrum Authentic
Acetyl Content acetyl content: < 4%
Viscosity 0.01 to 0.03 (sec.)
Avg. Mol. Wt. or Mol. Wt. Range 16000 (Mn)

Specifications

Chemical Name or Material Cellulose acetate butyrate
Name Note Butyryl Content 50-54%
CAS 9004-36-8
MDL Number MFCD00081513