missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cellulose Acetate Butyrate, Butyryl Content 50-54%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 404225000

 View more versions of this product

Catalog No. AC404225000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Cellulose acetate butyrate
Butyryl Content 50-54%
16000 (Mn)
Granular Crystalline Powder
hydroxyl content: 1.5% (typically)
Glass bottle
White
acetyl content: < 4%
9004-36-8
butyryl content: 50 to 54%
MFCD00081513
Authentic
0.01 to 0.03 (sec.)
500g

Chemical Identifiers

9004-36-8
MFCD00081513
Cellulose acetate butyrate, Butyryl Content 50-54%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA