missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cellulose acetate butyrate, butyryl content 16.5-19.0%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 177790050

 View more versions of this product

Catalog No. AC177790050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Fibrous Solid
White
butyryl content: 16.5 to 19.0%
Glass bottle
9004-36-8
3% max.
Authentic
5g
14 to 24 (sec.)
hydroxyl content: 1.5% (typically)
Cellulose acetate butyrate
MFCD00081513
acetyl content: 28.0 to 31.0%
approximate M.W.: 200000 (GPC)

Chemical Identifiers

9004-36-8
MFCD00081513
Cellulose acetate butyrate

Safety and Handling

SDS & Safety Documents

TSCA : TSCA