Please login to your online account to display your discounted pricing

Cellulose acetate butyrate, butyryl content 16.5-19.0%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cellulose acetate butyrate
CAS 9004-36-8
MDL Number MFCD00081513
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC177790050

  SDS
thermo fisher scientific
177790050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fibrous Solid
Quantity 5g
Color White
TSCA TSCA
Chemical Name or Material Cellulose acetate butyrate
Butyryl Content butyryl content: 16.5 to 19.0%
CAS 9004-36-8
Hydroxyl Content hydroxyl content: 1.5% (typically)
Packaging Glass bottle
Viscosity 14 to 24 (sec.)
Avg. Mol. Wt. or Mol. Wt. Range approximate M.W.: 200000 (GPC)
Acetyl Content acetyl content: 28.0 to 31.0%
MDL Number MFCD00081513
Infrared Spectrum Authentic
Loss on Drying 3% max.