Promega CellTiter-Glo™ 2.0 Assay DFS Item
The CellTiter-Glo™ 2.0 Assay provides a homogeneous method of determining the number of viable cells in culture by quantitating the amount of ATP present, which indicates the presence of metabolically active cells.
$226.37 - $3931.29
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG9243 DFS Item
View Documents Promotion Details
Promega
G9243
500 mL
Each for $3,931.29
PRG9242 DFS Item
View Documents Promotion Details
Promega
G9242
100 mL
Each for $925.25
PRG9241 DFS Item
View Documents Promotion Details
Promega
G9241
10 mL
Each for $226.37
Description
Description
  • A Luminescent Cell Viability Assay for Fast, Easy, Everyday Use
  • Reduced preparation time
  • Convenient repeat use over multiple days
  • Same great performance as classic CellTiter-Glo(R) Assay
SDS
missing translation for 'documents'
missing translation for 'documents'