missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One CELLSTAR High-Profile Cell Culture Flasks
Free of detectable DNase, RNase and human DNA
$330.09 - $474.50
Specifications
Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic |
---|---|
Closure Material | HDPE |
Capacity (Metric) | 650 mL |
Material | Polystyrene |
Neck Style | Canted |
Catalog Number | Mfr. No. | Surface Coating | Closure Type | Description | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Coating | Closure Type | Description | Price | Quantity | |||
07000381
|
Greiner Bio-One
661190 |
Hydrophobic surface (without treatment) | Screw Cap | Standard Suspension Culture Flask |
Case of 40 Each for $358.71
|
|
|||
07000383
|
Greiner Bio-One
661160 |
Physical Surface Treatment | Screw Cap | Standard Cell Culture Flask, TC treated |
Case of 40 Each for $330.09
|
|
|||
07000382
|
Greiner Bio-One
661175 |
Physical Surface Treatment | Filter Screw Cap | Standard Cell Culture Flask, TC treated |
Case of 40 Each for $474.50
|
|
|||
Description
- Design of flasks allows efficient access to the cell lawn with cell scrapers and optimal access with pipettes
- Graduation on the side
- Neck of the high profile flask reduces risk of wetting inside of cap with medium, and thus provides additional protection against contamination
- Filter screw cap
Specifications
Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic | |
650 mL | |
Canted | |
4 | |
Sterile |
HDPE | |
Polystyrene | |
40 | |
Yes |