Greiner Bio-One CELLMASTER™ Polystyrene Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
$437.97 - $852.31
Specifications
Sterility Sterile
Bottle Surface Smooth
Cell Growth Area 1700cm2
Material Polystyrene
Diameter (Metric) 122mm
View More Specs
Products 3
Catalog Number Mfr. No. No. per Pack No. per Case Price Quantity  
Catalog Number Mfr. No. No. per Pack No. per Case Price Quantity  
07000748 View Documents Greiner Bio-One
682060
1 12 Case of 12 Each for $444.40
07000744 View Documents Greiner Bio-One
682012
12 12 Case of 12 Each for $437.97
07000746 View Documents Greiner Bio-One
682024
12 24 Case of 24 Each for $852.31
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co
Specifications
Specifications
Sterile
1700cm2
122mm
Long Form
4.8 in.
500mm
Smooth
Polystyrene
USP Class VI end product testing
Cell cuture
Clear