Greiner Bio-One CELLMASTER™ Polystyrene Filter Cap Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene Filter Cap Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
$467.60 - $467.60
Specifications
Sterility Sterile
Bottle Surface Smooth
Cell Growth Area 1700cm2
Material Polystyrene
Diameter (Metric) 122mm
View More Specs
Products 2
Catalog Number Mfr. No. No. per Pack No. per Case Price Quantity  
Catalog Number Mfr. No. No. per Pack No. per Case Price Quantity  
07000745 View Documents Greiner Bio-One
682015
1 12 Case of 12 Each for $467.60
07000747 View Documents Greiner Bio-One™
682025
12 24 N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co
Specifications
Specifications
Sterile
1700cm2
122mm
Long Form
4.8 in.
500mm
Smooth
Polystyrene
USP Class VI end product testing
Cell cuture
Clear