Greiner Bio-One CELLMASTER™ Polystyrene 5XL Roller Bottles - Long Form
For use in cell culture. Greiner Bio-One CELLMASTER™ Polystyrene 5XL Roller Bottles - Long Form renders cell adhesion through physical surface treatment.
$1,104.37 - $2,036.06
Specifications
Surface Treatment TC Treated
Sterility Sterile
Bottle Surface Ribbed
Cell Growth Area 4250cm2
Material Polystyrene
View More Specs
Products 2
Catalog Number Mfr. No. No. per Pack No. per Case Price Quantity  
Catalog Number Mfr. No. No. per Pack No. per Case Price Quantity  
07000750 View Documents Greiner Bio-One
682070
1 12 Case of 12 Each for $1,104.37
07000751 View Documents Greiner Bio-One
682072
12 24 Case of 24 Each for $2,036.06
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co
Specifications
Specifications
TC Treated
Ribbed
Polystyrene
Long Form
4.8 in.
500mm
Sterile
4250cm2
122mm
Cell cuture
Clear