missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One CELLMASTER™ Polyethylene Terephthalate (PET) Roller Bottles - Short Form

For use in cell culture. Greiner Bio-One CELLMASTER™ Polyethylene Terephthalate (PET) Roller Bottles - Short Form is suitable for many adherent cell lines, is stable and gas permeable.

$291.80 - $444.31

Specifications

Sterility Sterile
Bottle Surface Smooth
Cell Growth Area 850cm2
Material Polystyrene
Height (English) 10.8 in.
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
07000733 View Documents Greiner Bio-One
680160
116 mm Case of 18 Each for $328.06
 
07000734 View Documents Greiner Bio-One
680180
122 mm Case of 18 Each for $291.80
 
07000345 View Documents Greiner Bio-One
680170
Case of 30 Each for $444.31
 
Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co
Specifications

Specifications

Sterile
850cm2
10.8 in.
USP Class VI end product testing
Cell cuture
Clear
Smooth
Polystyrene
1
Short form
4.5 in.
276mm