Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform DFS Item
Integrated system for high content analysis
Specifications
Light Source LED, solid-state 7-color light engine used with provided filter sets offers the following excitation/emission capabilities (Ex/Em, in nm): 386/440 (e.g., Alexa Fluor 350 dye); 438/480 (e.g., Alexa Fluor 405 dye); 485/521 (e.g., Alexa Fluor 488 dye); 549/600 (e.g., Alexa Fluor 555 dye); 560/607 (e.g., Alexa Fluor 594 dye); 650/694 (e.g., Alexa Fluor 647 dye); 740/810 (e.g., Alexa Fluor 750 dye)
Objectives 3-position objective turret (range from 2x–40x, low and high NA)
Camera Photometrics X1 camera High-resolution, thermoelectrically cooled to 0°C; 14-bit dynamic range; FOV 2208 x 2208 pixel array; 4.54 micron pixel size; 75% peak Quantum Efficiency (QE); USB 2.0 (no additional CameraLink or Firewire cards needed); Multiple binning options; Binning modes: 1x1, 2x2, or 4x4
View More Specs
Alert:
For research use only
Products 3
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
CX7A1110 View Documents Thermo Scientific™
CX7A1110
CellInsight™ CX7 High Content Analysis Platform N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CX7C1115 View Documents Thermo Scientific™
CX7C1115
CellInsight™ CX7 High Content Analysis Platform and Store Standard Edition plus Robotic Plate Handling N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CX7B1112 View Documents Thermo Scientific™
CX7B1112
CellInsight™ CX7 High Content Analysis Platform and Store Standard Edition N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Integrated system for high content analysis
The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.
The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.
Features of the CellInsight CX7 HCA Platform
  • 7-channel fluorescent imaging
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured for fully automated plate handling and scanning
Capabilities
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Use HCS Studio software for integrated instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Compatible with Molecular Probes™ fluorescent detection reagents
Hardware and Software
  • Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
  • Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler
Specifications
Specifications
LED, solid-state 7-color light engine used with provided filter sets offers the following excitation/emission capabilities (Ex/Em, in nm): 386/440 (e.g., Alexa Fluor 350 dye); 438/480 (e.g., Alexa Fluor 405 dye); 485/521 (e.g., Alexa Fluor 488 dye); 549/600 (e.g., Alexa Fluor 555 dye); 560/607 (e.g., Alexa Fluor 594 dye); 650/694 (e.g., Alexa Fluor 647 dye); 740/810 (e.g., Alexa Fluor 750 dye)
3-position objective turret (range from 2x–40x, low and high NA)
Photometrics X1 camera High-resolution, thermoelectrically cooled to 0°C; 14-bit dynamic range; FOV 2208 x 2208 pixel array; 4.54 micron pixel size; 75% peak Quantum Efficiency (QE); USB 2.0 (no additional CameraLink or Firewire cards needed); Multiple binning options; Binning modes: 1x1, 2x2, or 4x4