Please login to your online account to display your discounted pricing

Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform DFS Item


Compact, affordable high content screening

Manufacturer: thermo scientific™  CX51114S

 View more versions of this product

Catalog No. CX51114S

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Weight (English) 99.2 lb.
Light Source LED, solid-state five-color light engine used with provided filter sets offers the following  excitation/emission capabilities:  Ex 386/Em 440 (e.g., Hoechst); Ex 485/Em 521 (e.g., Alexa Fluor 488); Ex 560/Em 607 (e.g., TRITC); Ex 650/Em 694 (e.g., Cy5); Ex 740/Em 810 (e.g., Alexa Fluor 750)
Depth (English) 23.4 in.
Description CeLLInsight™ CX5 High Content Screening Platform with Store™ Standard Edition (SE) and Orbitor™ RS Plate Mover (Stacker Configuration)
Weight (Metric) 45kg
Height (English) 14.6 in.
Height (Metric) 37.2cm
Width (Metric) 41.4cm
Width (English) 16.3 in.
Depth (Metric) 59.4cm
Camera Photometrics X1 CCD: High-resolution, thermoelectrically cooled to 0°C; 14-bit dynamic range; FOV 2208 x 2208 pixel array; 4.54 micron pixel size; 75% peak Quantum Efficiency (QE); USB 2.0 (no additional CameraLink or Firewire Cards needed)
Wattage 300w
Objectives Standard: 10X (0.30NA); 20X (0.4NA) Olympus objectives
Optional: 2x (0.08NA); 4x (0.16NA) Olympus objectives
PC Windows™ 7 64-bit PC with minimum quad core 3.7GHz processor, 8GB RAM, and 500GB Hard Drive
Software Thermo Scientific™ HCS Studio™ 2.0 Software, Thermo Scientific™ Store™ Image and Database Management Software
Data Management Compatible with Thermo Scientific™ Store™Image and Database Management Software

Rapidly perform high content imaging assays with the affordable, compact Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform.

The CellInsight CX5 High Content Screening (HCS) Platform is a fast, simple-to operate, automated cellular imaging and analysis benchtop platform designed for quantitative imaging. For the researcher looking to gain the advantage of quantitative cell analysis for the first time or for those who are looking to increase scale and capacity, the CellInsight CX5 HCS Platform offers a costs effective, but powerful way to quickly automate quantitative cellular imaging.

Highlights:
  • Flexible – designed to work with or without fluorescent labeling, the new CellInsight CX5 HCS Platform uses transmitted light allowing users to explore more cell biology without the restriction of fluorescent dyes or to use the full capabilities of a five-color light engine
  • Affordable – automated high content imaging for all budgets
  • Easy to Use – quickly build and optimize image-based assays with included Thermo Scientific™HCS Studio™ 2.0 Cell Analysis Software
  • Performance – scan and analyze a 96-well plate in under four minutes
  • Quality – high-quality imaging and virtually maintenance-free operation
  • Compact – fits on a bench and is less than 2 ft. on all sides
Capabilities:
  • Make individual cell, subcellular and multicellular measurements
  • Save time by automatically collecting only the images and data you need to be statistically relevant
  • Make multiple measurements per object, over time if desired
  • Measure subpopulations and reject undesired objects automatically
Easy to Use Applications:
  • Comprehensively pre-validated and user configurable image analysis without scripting or programming
  • Exploratory assay development tools and specific biology optimized tools
  • Define biological thresholds for outputs based on control populations
  • Ability to create multi-parameter “gates” from individual output measurements
Hardware and Software:
  • HCS Studio 2.0 Cell Analysis Software – the premier high content instrument control, visualization and analysis program for novice to advanced users featuring image/data portability
  • Ultra-stable, long-life light source reduces variability, increases data quality, and decreases operating costs
  • Fluorescence options which include near-IR wavelengths
  • Proprietary Thermo Scientific™ AccuFocus System is fast, yet flexible, imaging cells with high-fidelity every time
  • Unique intelligent acQuisition (iQ) real time analysis feedback to optimize image collection and data quality
  • Compatible with SBS standard microplates (6-1536 well) and slides
Compatible with: Optional Automation – Thermo Scientific™ Orbitor™ RS Plate Mover and other Thermo Scientific™ Laboratory Automation Solutions; Optional Modules - Thermo Scientific™ High Content Modules

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.