Please login to your online account to display your discounted pricing

Cell Size and Diffusion Lab


A quick, easy and safe visual demonstration of cell size and diffusion.

Manufacturer: fisher science education  AZ-20-1013

 View more versions of this product

Catalog No. S63067

  • / Each
 

Description & Specifications

Specifications

Time Range 30 minutes
For Use With (Application) Easy and safe visual demonstration of cell size & diffusion
Product Type Why Cells Aren'T Big Kit
Includes Agar, diffusion media, cups, gloves and rulers, dilute base, comprehensive teacher and student guides
Kit Size 40 students
Class Size 40 students