Thermo Scientific™ Cell Roller System Accessories for Environmental Chambers and Incubators
Allow production of monolayer cell cultures in standard roller culture vessels.
$2,296.83 - $3,660.00
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Installation Product Type Price Quantity  
Catalog Number Mfr. No. Description Installation Product Type Price Quantity  
15-465-185 Thermo Scientific™
FOR500182
Reinforced Floor with Removable Ramp Customer installed Reinforced Floor Options Each for $2,296.83
19-004-9 Thermo Scientific™
PSI190049
Add-On Tier (5 positions) Customer installed Cell Roller System Each for $3,660.00
19-077-7 Thermo Scientific™
PSI190777
Reinforced Floor with Removable Ramp (extends 23 Factory installed Reinforced Floor Options Each for $2,595.17
22-807-7 Thermo Scientific™
PSI228077
Alarm system for Cell Rollers Factory installed Cell Roller System Each for $3,112.96
Specifications
Description
Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators. Oxygenation and exposure of the cells to the media growth area are improved. Culture yields are increased by the uniform temperature control and cell roll system’s continuous, gentle rotation.