Cell Locker™ Chamber
Protected chambers for your most sensitive cells
$1,883.77 - $1,951.04
Products 2
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
11675972 View Documents Thermo Scientific
50151650
Cell Locker Chamber with stainless steel tray Each for $1,883.77
11675973 View Documents Thermo Scientific
50154739
Cell Locker Chamber with copper tray Each for $1,951.04
Description
Description
Individual chambers maximize security against costly cross contamination. Each Cell Locker Chamber has dual 0.2μm membrane filters that permit air circulation but exclude microbial contaminants. Independent tests demonstrate that microorganisms cannot pass between closed chambers. Individual Cell Locker Chambers serve to quarantine cell types or different projects, offering enhanced protection for valuable cultures.
  • Each individual Cell Locker Chamber is effectively a quarantine chamber.
  • Dual 0.2μm membrane filters permits air circulation but exclude microbial contaminants.
  • The replacable 0.2µm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity.
  • Independent 3rd party tests validate protection against circulating microorganisms (when used as directed)
Each Cell Locker Chamber holds 9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96-well plates.
  • Dishes and flasks can be removed individually on a work tray or inside the entire Cell Locker Chamber with optional transport cover, thus protecting samples from outside air.
  • The non skid tray slides out to allow access to culture vessels, or the entire tray can be removed.
  • The Cell Locker Chamber can be manually disinfected or can be autoclaved.
Warranty: 1 year