Please login to your online account to display your discounted pricing
GE Healthcare Cell Holder
Manufacturer: ge healthcare  80210613
 View more versions of this product
Catalog No. BC80-2106-13
  • / Each
Description & Specifications
Specifications
Item Description Cell Holder
Compatibility Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300
For Use With Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300
An adjustable single cell holder for 10-40mm pathlength cells. Operational temperature range from ambient to 100°C and compatible with Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300 spectrophotometer models. Suitable for enzyme kinetics studies that need to be under thermostatic control from a circulation bath. This water-heated cell holder is an alternative to the Peltier heated versions.
  • Adjustable single cell holder for 10 to 40mm pathlength cells.
  • Temperature range from ambient to 100°C.
  • Compatible with Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 models.