Please login to your online account to display your discounted pricing
GE Healthcare Cell Holder
$605.62 - $5,485.11
Products
Catalog Number Mfr. No. For Use With Price Quantity    
BC80-2106-05
ge healthcare
80210605
Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 spectrophotometers Each for $605.62
BC80-2106-08
ge healthcare
80210608
Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300 spectrophotometer models Each for $2,621.41
BC80-2106-09
ge healthcare
80210609
Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 models. Each for $1,045.49
BC80-2106-13
ge healthcare
80210613
Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300 Each for $5,485.11
BC80210714
ge healthcare
80210714
Ultrospec Each for $1,453.35
Description & Specifications
An adjustable single cell holder for 10-40mm pathlength cells. Operational temperature range from ambient to 100°C and compatible with Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300 spectrophotometer models. Suitable for enzyme kinetics studies that need to be under thermostatic control from a circulation bath. This water-heated cell holder is an alternative to the Peltier heated versions.
  • Adjustable single cell holder for 10 to 40mm pathlength cells.
  • Temperature range from ambient to 100°C.
  • Compatible with Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 models.