Biochrom Micro-Volume Cell Holder
Micro-Volume Cell Holder is for use with 50 μL cuvettes. Accessory for the Ultrospec 7500 and 2100 Pro Spectrophotometer.
Supplier: Biochrom 80210609
Description
- Micro-volume cell holder
- For the Ultrospec 2100 Pro Spectrophotometer
- Use with 50 μL cuvette (80-2076-38)
Specifications
Ultrospec 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models | |
Cell Holder | |
Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models |
Cell Holder | |
Reusable | |
Round |