missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ Cell Culture Flasks with Vented Filter Cap, Non-Treated

Preferred vessel for long term cultivation or large-scale expansion of cells, providing protection against contamination. Eppendorf™ Cell Culture Flasks with Vented Filter Cap, Non-Treated are sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free and non-cytotoxic.

Manufacturer:  Eppendorf™ 0030711025

 View more versions of this product

Catalog No. 13-690-058



Description

Description

  • Features ConvexAcces™ geometry for facilitated access to growth area and easier and safe cell treatment
  • Optimized gas exchange and protection against contamination through high efficiency air filter technology
  • Anti-rolling cap with corrugation facilitates cultivation steps when cap needs to be set aside
  • 100% in-line control for leakage-free flasks and maximum safety
  • Direct surface identification on flask and cap
  • Available in three sizes: T–25, T–75 and T–175 with non-treated surfaces
Specifications

Specifications

No
Vented filter cap
77.9cm2
Polystyrene
T-75 Cell culture flask
Non-treated
8.0 to 20.0mL
35.2mm
Transparent
20.0mL
  • Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene
  • Performance of cell-based assays and microscopic analysis
SAL 10-6
−86 °C to +60 °C
47.0g
Documents