Please login to your online account to display your discounted pricing

Celite 545 Filter Aid (Powder), Fisher Chemical


Not acid-washed

Quantity 50 lb.
Packaging Fiber Drum

Manufacturer: thermo fisher scientific  FLC21250LB

 View more versions of this product

Catalog No. C212-50LB

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50 lb.
Physical Form Solid
Density 2.3g/cm³
Color White
pH 9 to 10.5
Packaging Fiber Drum
MDL Number MFCD00132803
CAS 68855-54-9
Synonym Diatomaceous Earth
Identification Pass Test
Chemical Name or Material Celite™ 545 Filter Aid
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Celite™ 545 Filter Aid
CAS 68855-54-9
Physical Form Solid
MDL Number MFCD00132803
Synonym Diatomaceous Earth
Accessories
 

SDS & Safety Documents