missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Celite 545 Filter Aid (Powder), Fisher Chemical™
Not acid-washed
Supplier: Fisher Chemical FLC212500
Specifications
Solid | |
2.3 g/cm3 | |
White | |
68855-54-9 | |
CNa2O3 | |
Diatomaceous Earth, Kieselguhr | |
C(=O)([O-])[O-].[Na+].[Na+] | |
105.988 | |
CHEBI:29377 |
500 g | |
9 to 10.5 | |
Celite™ 545 Filter Aid | |
Poly Bottle | |
MFCD00132803 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 |
Chemical Identifiers
68855-54-9 | |
105.988 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
disodium;carbonate |
CNa2O3 | |
MFCD00132803 | |
Diatomaceous Earth, Kieselguhr | |
CHEBI:29377 | |
C(=O)([O-])[O-].[Na+].[Na+] |