missing translation for 'onlineSavingsMsg'
Learn More

Celite 545 Filter Aid (Powder), Fisher Chemical™

Catalog No. p-31544
Change view
Click to view available options
Quantity:
500 g
25 lb.
50 lb.
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C212500 500 g Poly Bottle
C21225LB 25 lb. Fiber Drum
C21250LB 50 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C212500 Supplier Thermo Fisher Scientific Supplier No. FLC212500
Only null left

Not acid-washed

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym Diatomaceous Earth, Kieselguhr
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Physical Form Solid
Density 2.3 g/cm3
Quantity 500 g
pH 9 to 10.5
Color White
Packaging Poly Bottle
Chemical Name or Material Celite™ 545 Filter Aid