missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Celite™ 545 Filter Aid, Flux-Calcined, Honeywell Fluka™
Filter aid, treated with sodium carbonate, flux calcined
$236.35 - $799.23
Chemical Identifiers
CAS | 68855-54-9 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.988 |
MDL Number | MFCD00132803 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
IUPAC Name | disodium;carbonate |
SMILES | C(=O)([O-])[O-].[Na+].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
68855-54-9 | |
105.988 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
CHEBI:29377 | |
C(=O)([O-])[O-].[Na+].[Na+] |
CNa2O3 | |
MFCD00132803 | |
10340 | |
disodium;carbonate |
Specifications
1 kg | |
CNa2O3 | |
NONH for all modes of transport | |
C(=O)([O-])[O-].[Na+].[Na+] | |
105.988 | |
CHEBI:29377 |
68855-54-9 | |
MFCD00132803 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
Diatomaceous Earth, Flux-Calcined, Treated with sodium carbonate filter aid |
Safety and Handling
P260
H373
EINECSNumber : 272-489-0