missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Celestine Blue, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 195560100

 View more versions of this product

Catalog No. AC195560100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Celestine blue
C17H18ClN3O4
15, 1961
Authentic
7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride
54684697
363.79
Glass bottle
Complies
10 g
1562-90-9
MFCD00011927
OJAMERVLRXYMPD-UHFFFAOYSA-N
CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-]
363.798
638 to 648nm
Extra Pure
700 min. (in water)
Black to Brown
Powder

Chemical Identifiers

1562-90-9
363.798
OJAMERVLRXYMPD-UHFFFAOYSA-N
7-(diethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxamide;chloride
C17H18ClN3O4
MFCD00011927
54684697
CCN(CC)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)N.[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only