Please login to your online account to display your discounted pricing

Celecoxib, Tocris™

Selective cyclooxygenase-2 (COX-2) inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Celecoxib
Assay Percent Range >99%
Molecular Formula C17H14F3N3O2S
Formula Weight 381.37
Synonym 4-[5-(4-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

378610

 
r & d systems
RDS378610
10mg Each for $201.36

378650

 
r & d systems
RDS378650
50mg Each for $844.73
Description & Specifications

Specifications

Chemical Name or Material Celecoxib
Quantity 10mg
CAS 169590-42-5
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C17H14F3N3O2S
Formula Weight 381.37
Synonym 4-[5-(4-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide
Safety and Handling

Store at Room Temperature