Please login to your online account to display your discounted pricing

Alfa Aesar™ Ceftizoxime sodium, 95%

Chemical Identifiers

Chemical Name or Material Ceftizoxime sodium
CAS 68401-82-1
Formula Weight 405.38
MDL Number MFCD01682036
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66554MF

 
alfa aesar™
J66554MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 405.38
Chemical Name or Material Ceftizoxime sodium
Physical Form Powder
Assay 95%
CAS 68401-82-1
MDL Number MFCD01682036
Solubility Information Soluble in water
Color White to pale yellow
Melting Point 227°C
Storage Note 1 2° to 8°C
Quantity 50mg
RTECS Number XI0368000

  • A broad spectrum antibiotic targeting a wide variety of gram positive and gram negative bacteria

Safety and Handling

2° to 8°C