Please login to your online account to display your discounted pricing

Alfa Aesar™ Ceftibuten, 95%

A broad spectrum antibiotic

Chemical Identifiers

Chemical Name or Material Ceftibuten
CAS 97519-39-6
Formula Weight 410.42
MDL Number MFCD00864918
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66807MA

 
alfa aesar™
J66807MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66807MC

 
alfa aesar™
J66807MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66807MF

 
alfa aesar™
J66807MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 410.42
Chemical Name or Material Ceftibuten
Physical Form Powder
Assay 95%
MDL Number MFCD00864918
CAS 97519-39-6
Solubility Information Soluble in aqueous solutions; Also soluble in DMSO
Color White to pale brown
RTECS Number XI0367220
Boiling Point 966°C
Storage Note 1 2° to 8°C
Quantity 10mg

Applications
Ceftibuten is used to treat acute bacterial exacerbations of chronic bronchitis (ABECB), acute bacterial otitis media, pharyngitis, andtonsilitis. It is also indicated for pneumonia, infections of the urinary tract, enteritis, and gastroenteritis.

Solubility
Soluble in aqueous solutions. Also soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

2° to 8°C