Please login to your online account to display your discounted pricing

Alfa Aesar™ Ceftibuten, 95%


A broad spectrum antibiotic

Quantity 100mg

Manufacturer: alfa aesar™  J66807MC

 View more versions of this product

Catalog No. AAJ66807MC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 410.42
Chemical Name or Material Ceftibuten
Assay 95%
MDL Number MFCD00864918
CAS 97519-39-6
Physical Form Powder
Solubility Information Soluble in aqueous solutions; Also soluble in DMSO
Boiling Point 966°C
Color White to pale brown
Storage Note 1 2° to 8°C
Quantity 100mg
RTECS Number XI0367220

Specifications

Chemical Name or Material Ceftibuten
CAS 97519-39-6
Formula Weight 410.42
MDL Number MFCD00864918

Applications
Ceftibuten is used to treat acute bacterial exacerbations of chronic bronchitis (ABECB), acute bacterial otitis media, pharyngitis, andtonsilitis. It is also indicated for pneumonia, infections of the urinary tract, enteritis, and gastroenteritis.

Solubility
Soluble in aqueous solutions. Also soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Accessories

2° to 8°C

SDS & Safety Documents