missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ceftazidime pentahydrate, 95%

Manufacturer:  Alfa Aesar™ J6646003

 View more versions of this product

Catalog No. AAJ6646003



Description

Description

Solubility
Sparingly soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications

Specifications

Ceftazidime pentahydrate
C22H32N6O12S2
Sparingly soluble in aqueous solution
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
636.648
CHEBI:3509
Powder
White
78439-06-2
MFCD00153936
NMVPEQXCMGEDNH-TZVUEUGBSA-N
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
131673973
636.65
95%
1g

Chemical Identifiers

78439-06-2
636.648
NMVPEQXCMGEDNH-TZVUEUGBSA-N
CHEBI:3509
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
C22H32N6O12S2
MFCD00153936
131673973
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P333+P313-P342+P311-P363-P501c

H317-H319-H334

RTECSNumber : UU2230000

TSCA : No

Recommended Storage : Keep cold

Documents