Please login to your online account to display your discounted pricing

Alfa Aesar™ Cefradine

$78.66 - $78.66

Chemical Identifiers

Chemical Name or Material Cefradine
CAS 38821-53-3
Molecular Formula C16H19N3O4S
Formula Weight 349.4
MDL Number MFCD00865048
Merck Index 14,1986
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6649603

  SDS
alfa aesar™
J6649603
1g Each for $78.66

AAJ6649606

  SDS
alfa aesar™
J6649606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ6649614

  SDS
alfa aesar™
J6649614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 349.4
Storage Note 1 2° to 8°C
CAS 38821-53-3
MDL Number MFCD00865048
Chemical Name or Material Cefradine
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,1986
Physical Form Powder
Recommended Storage 2° to 8°C
Color White to Pale Yellow
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 1 Danger
Boiling Point 898°C
Melting Point 140° to 142°C
RTECS Number XI0336000
Quantity 1g
Solubility Information Soluble in water
Molecular Formula C16H19N3O4S

  • A semi-synthetic cephalosporin antibiotic that is used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

2° to 8°C

2° to 8°C